Gene Transcripts
- Gene Symbol
- Anxa1
- Entrez Gene ID
- 25380
- Gene Name
- annexin A1
- Gene Aliases
- Anx1, p35
- Chromosome Location
- Chr. 1: 237893983 - 237921165 on Build Rnor_6.0
- UniGene ID
- Rn.1792
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012904.2 |
NP_037036.1
|
7, 8 |
624 |
|
XM_006231159.1 |
XP_006231221.1
|
8, 9 |
792 |
|
XM_008760295.1 |
XP_008758517.1
|
7, 8 |
711 |
|
GenBank mRNA |
BC061710.1 |
- |
7, 8 |
622 |
|
FQ213489.1 |
- |
7, 8 |
639 |
|
FQ215033.1 |
- |
7, 8 |
639 |
|
FQ216134.1 |
- |
7, 8 |
639 |
|
FQ228716.1 |
- |
7, 8 |
640 |
|
FQ229071.1 |
- |
7, 8 |
640 |
|
FQ229198.1 |
- |
6, 7 |
560 |
|
M19967.1 |
- |
6, 7 |
591 |
|
Y00446.1 |
- |
7, 8 |
625 |
|
View Full Product Details