Gene Transcripts
- Gene Symbol
- Ntf4
- Entrez Gene ID
- 25730
- Gene Name
- neurotrophin 4
- Gene Aliases
- NT4P, Ntf5
- Chromosome Location
- Chr. 1: 101405314 - 101408145 on Build Rnor_6.0
- UniGene ID
- Rn.44225
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013184.3 |
NP_037316.2
|
3 |
716 |
|
GenBank mRNA |
S69323.1 |
- |
3 |
716 |
|
View Full Product Details