Gene Transcripts
- Gene Symbol
- SLC25A1
- Entrez Gene ID
- 6576
- Gene Name
- solute carrier family 25 member 1
- Gene Aliases
- CTP, D2L2AD, SEA, SLC20A3
- Chromosome Location
- Chr. 22: 19175575 - 19178863 on Build GRCh38
- UniGene ID
- Hs.111024
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256534.1 |
NP_001243463.1
|
2, 3 |
481 |
|
NM_001287387.1 |
NP_001274316.1
|
3, 4 |
335 |
|
NM_005984.4 |
NP_005975.1
|
3, 4 |
500 |
|
GenBank mRNA |
AK292313.1 |
- |
3, 4 |
404 |
|
AK298204.1 |
- |
3, 4 |
333 |
|
BC004980.1 |
- |
3, 4 |
361 |
2957917 |
BC008061.2 |
- |
3, 4 |
363 |
2958945 |
BC018590.1 |
- |
3, 4 |
258 |
|
BG473148.1 |
- |
2, 3 |
455 |
|
BP265446.1 |
- |
3, 4 |
335 |
|
DR006840.1 |
- |
3, 4 |
402 |
|
HM037273.1 |
- |
2, 3 |
336 |
|
L75823.1 |
- |
3, 4 |
409 |
|
L77567.1 |
- |
3, 4 |
382 |
|
U25147.1 |
- |
3, 4 |
384 |
|
View Full Product Details