Gene Transcripts
- Gene Symbol
- Hcn3
- Entrez Gene ID
- 114245
- Gene Name
- hyperpolarization-activated cyclic nucleotide-gated potassium channel 3
- Gene Aliases
- Chromosome Location
- Chr. 2: 188458851 - 188471916 on Build Rnor_6.0
- UniGene ID
- Rn.48801
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053685.1 |
NP_446137.1
|
7, 8 |
1640 |
|
GenBank mRNA |
AF247452.1 |
- |
7, 8 |
1640 |
|
View Full Product Details