Gene Transcripts

Gene Symbol
Med11
Entrez Gene ID
287456
Gene Name
mediator complex subunit 11
Gene Aliases
RGD1563202
Chromosome Location
Chr. 10: 57057272 - 57059415 on Build Rnor_6.0
UniGene ID
Rn.19345
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105799.1 NP_001099269.1 3 343
XM_006246660.2 XP_006246722.1 3 681
GenBank mRNA FQ221563.1 - 3 355

View Full Product Details