Gene Transcripts

Gene Symbol
Wdr83
Entrez Gene ID
288924
Gene Name
WD repeat domain 83
Gene Aliases
Morg1, RGD1306947
Chromosome Location
Chr. 19: 26188644 - 26194193 on Build Rnor_6.0
UniGene ID
Rn.3570
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047847.1 NP_001041312.1 7 718
GenBank mRNA AY940050.1 - 7 718

View Full Product Details