Gene Transcripts

Gene Symbol
Mettl13
Entrez Gene ID
289159
Gene Name
methyltransferase like 13
Gene Aliases
RGD1311526
Chromosome Location
Chr. 13: 80427595 - 80441352 on Build Rnor_6.0
UniGene ID
Rn.18927
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134501.1 NP_001127973.1 8 2336
XM_006250152.2 XP_006250214.1 9 2264
XM_008769622.1 XP_008767844.1 8 2366

View Full Product Details