Gene Transcripts

Gene Symbol
Spcs2
Entrez Gene ID
293142
Gene Name
signal peptidase complex subunit 2 homolog (S. cerevisiae)
Gene Aliases
RGD1311253
Chromosome Location
Chr. 1: 164829896 - 164849749 on Build Rnor_6.0
UniGene ID
Rn.7939
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191601.1 NP_001178530.1 4, 5 502
GenBank mRNA FQ214686.1 - 4, 5 504
FQ232393.1 - 1, 2 95
FQ233613.1 - 1, 2 95

View Full Product Details