Gene Transcripts

Gene Symbol
Sytl1
Entrez Gene ID
297872
Gene Name
synaptotagmin-like 1
Gene Aliases
Slp-1
Chromosome Location
Chr. 5: 151448639 - 151459875 on Build Rnor_6.0
UniGene ID
Rn.16862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025651.1 NP_001020822.1 9, 10 1011
XM_006239013.2 XP_006239075.1 9, 10 1849
GenBank mRNA BC098679.1 - 9, 10 1011

View Full Product Details