Gene Transcripts

Gene Symbol
Ndufb6
Entrez Gene ID
297990
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6
Gene Aliases
Chromosome Location
Chr. 5: 56567109 - 56576676 on Build Rnor_6.0
UniGene ID
Rn.104528
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106646.1 NP_001100116.1 2 348
GenBank mRNA FQ216998.1 - 2 275

View Full Product Details