Gene Transcripts

Gene Symbol
Mageb5
Entrez Gene ID
302442
Gene Name
melanoma antigen family B, 5
Gene Aliases
Mageb17, RGD1563078
Chromosome Location
Chr. X: 60849005 - 60852583 on Build Rnor_6.0
UniGene ID
Rn.218663
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106946.1 NP_001100416.1 2 1144
XM_006257012.2 XP_006257074.1 2 1118
XM_006257013.2 XP_006257075.1 2 1122

View Full Product Details