Gene Transcripts

Gene Symbol
Mrvi1
Entrez Gene ID
308899
Gene Name
murine retrovirus integration site 1 homolog
Gene Aliases
Chromosome Location
Chr. 1: 175681958 - 175796115 on Build Rnor_6.0
UniGene ID
Rn.33889
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105210.1 NP_001098680.1 15 2323
NM_001105211.1 NP_001098681.1 14 2076
XM_006230047.2 XP_006230109.1 15 2321
XM_006230048.2 XP_006230110.1 16 2507
XM_008759727.1 XP_008757949.1 15 2423
XM_008759728.1 XP_008757950.1 15 2642
XM_008759729.1 XP_008757951.1 16 2104

View Full Product Details