Gene Transcripts

Gene Symbol
Ell2
Entrez Gene ID
309918
Gene Name
elongation factor RNA polymerase II 2
Gene Aliases
Chromosome Location
Chr. 2: 2456189 - 2525590 on Build Rnor_6.0
UniGene ID
Rn.105837
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760603.1 XP_008758825.1 11 2516
XM_008760604.1 XP_008758826.1 11 1802
GenBank mRNA BC111075.1 - 2 128
FQ230057.1 - 4 510

View Full Product Details