Gene Transcripts

Gene Symbol
Noxa1
Entrez Gene ID
311793
Gene Name
NADPH oxidase activator 1
Gene Aliases
Chromosome Location
Chr. 3: 2299688 - 2309466 on Build Rnor_6.0
UniGene ID
Rn.162651
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100171.1 NP_001093641.1 5 571
XM_006233600.2 XP_006233662.1 5 858

View Full Product Details