Gene Transcripts

Gene Symbol
Ndufaf2
Entrez Gene ID
361894
Gene Name
NADH dehydrogenase (ubiquinone) complex I, assembly factor 2
Gene Aliases
Ndufa12l, RGD1560158
Chromosome Location
Chr. 2: 39322145 - 39434537 on Build Rnor_6.0
UniGene ID
Rn.103613
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_342191.5 XP_342192.1 4 437
GenBank mRNA FQ224641.1 - 4 395

View Full Product Details