Gene Transcripts

Gene Symbol
Mrgprx2
Entrez Gene ID
404640
Gene Name
MAS-related GPR, member X2
Gene Aliases
Mgrg6, MrgB1, Mrgprb1
Chromosome Location
Chr. 1: 103744699 - 103745583 on Build Rnor_6.0
UniGene ID
Rn.215681
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002280.1 NP_001002280.1 1 445

View Full Product Details