Gene Transcripts

Gene Symbol
Marveld3
Entrez Gene ID
498950
Gene Name
MARVEL domain containing 3
Gene Aliases
RGD1562056
Chromosome Location
Chr. 19: 41710076 - 41724923 on Build Rnor_6.0
UniGene ID
Rn.102273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109132.1 NP_001102602.1 2 505
XM_006255598.2 XP_006255660.1 2 531

View Full Product Details