Gene Transcripts

Gene Symbol
Mmrn1
Entrez Gene ID
500152
Gene Name
multimerin 1
Gene Aliases
Chromosome Location
Chr. 4: 90977534 - 91038747 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001071128.5 XP_001071128.2 7 3034
XM_006236596.2 XP_006236658.1 7 3034
XM_006236597.2 XP_006236659.1 8 3032

View Full Product Details