Gene Transcripts

Gene Symbol
Ndufa4
Entrez Gene ID
681024
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4
Gene Aliases
Chromosome Location
Chr. 4: 38233680 - 38240848 on Build Rnor_6.0
UniGene ID
Rn.793
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127684.2 NP_001121156.1 3, 4 308
GenBank mRNA BC167115.1 - 3, 4 307
FM055502.1 - 3, 4 204
FM088467.1 - 3, 4 288
FQ210865.1 - 3, 4 263
FQ217628.1 - 3, 4 251
FQ224471.1 - 3, 4 253
FQ224731.1 - 3, 4 279

View Full Product Details