Gene Transcripts

Gene Symbol
Mrpl39
Entrez Gene ID
684304
Gene Name
mitochondrial ribosomal protein L39
Gene Aliases
Chromosome Location
Chr. 11: 24181605 - 24200019 on Build Rnor_6.0
UniGene ID
Rn.216497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768604.1 XP_008766826.1 7, 8 1009
GenBank mRNA FQ225027.1 - 7, 8 968
FQ230552.1 - 7, 8 969

View Full Product Details