Gene Transcripts

Gene Symbol
Chmp1b
Entrez Gene ID
689364
Gene Name
charged multivesicular body protein 1B
Gene Aliases
Chromosome Location
Chr. 18: 62923620 - 62926150 on Build Rnor_6.0
UniGene ID
Rn.39132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109533.1 NP_001103003.1 1 1577
GenBank mRNA BC168175.1 - 1 1538

View Full Product Details