Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013019.2 | NP_001013037.1 | 2 | 455 | |
GenBank mRNA | AK087615.1 | - | 456 | ||
AK158268.1 | - | 2 | 393 | ||
BC107244.2 | - | 2 | 428 | 40052213 | |
BC107245.2 | - | 2 | 428 | 40052216 |