Gene Transcripts

Gene Symbol
2010107E04Rik
Entrez Gene ID
70257
Gene Name
RIKEN cDNA 2010107E04 gene
Gene Aliases
9430003J03Rik, AU043134, AV124504, Mp68
Chromosome Location
Chr. 12: 111961375 - 111966977 on Build GRCm38
UniGene ID
Mm.318
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027360.2 NP_081636.1 4 282
GenBank mRNA AK008333.1 - 4 283
BC055712.1 - 4 261 6408438
BC059719.1 - 4 270 6774233

View Full Product Details