Gene Transcripts

Gene Symbol
POMT1
Entrez Gene ID
10585
Gene Name
protein O-mannosyltransferase 1
Gene Aliases
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
Chromosome Location
Chr. 9: 131502895 - 131523806 on Build GRCh38
UniGene ID
Hs.522449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077365.1 NP_001070833.1 5 616
NM_001077366.1 NP_001070834.1 4 464
NM_001136113.1 NP_001129585.1 5 856
NM_001136114.1 NP_001129586.1 3 357
NM_007171.3 NP_009102.3 5 616
XM_005272156.1 XP_005272213.1 5 742
XM_005272158.1 XP_005272215.1 4 471
XM_005272159.1 XP_005272216.1 3 361
XM_005272162.2 XP_005272219.1 5 590
XM_006716932.1 XP_006716995.1 4 394
XM_017014203.1 XP_016869692.1 5 1064
XM_017014204.1 XP_016869693.1 4 399
XM_017014205.1 XP_016869694.1 5 538
GenBank mRNA AB209430.1 - 2 1588
AF095136.1 - 5 593
AK074874.1 - 5 574
AK074888.1 - 4 448
AK293529.1 - 3 269
AK295561.1 - 3 361
AK300393.1 - 4 484
AK301486.1 - 5 598
AK307876.1 - 5 574
AK315218.1 - 5 551
BC022877.2 - 5 510 4826806
BC065268.1 - 5 801 6155142

View Full Product Details