Gene Transcripts
- Gene Symbol
- NEK1
- Entrez Gene ID
- 4750
- Gene Name
- NIMA related kinase 1
- Gene Aliases
- NY-REN-55, SRPS2, SRPS2A, SRTD6
- Chromosome Location
- Chr. 4: 169393270 - 169612629 on Build GRCh38
- UniGene ID
- Hs.481181
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199397.1 |
NP_001186326.1
|
24, 25 |
2725 |
|
NM_001199398.1 |
NP_001186327.1
|
23, 24 |
2593 |
|
NM_001199399.1 |
NP_001186328.1
|
21, 22 |
2434 |
|
NM_001199400.1 |
NP_001186329.1
|
21, 22 |
2587 |
|
NM_012224.2 |
NP_036356.1
|
22, 23 |
2719 |
|
XM_006714228.1 |
XP_006714291.1
|
24, 25 |
2727 |
|
XM_011532003.1 |
XP_011530305.1
|
23, 24 |
2643 |
|
XM_011532004.1 |
XP_011530306.1
|
22, 23 |
2511 |
|
XM_017008249.1 |
XP_016863738.1
|
23, 24 |
2629 |
|
XM_017008250.1 |
XP_016863739.1
|
22, 23 |
2747 |
|
XM_017008251.1 |
XP_016863740.1
|
22, 23 |
2545 |
|
XM_017008252.1 |
XP_016863741.1
|
20, 21 |
2565 |
|
XM_017008253.1 |
XP_016863742.1
|
23, 24 |
2575 |
|
XM_017008254.1 |
XP_016863743.1
|
16, 17 |
1685 |
|
GenBank mRNA |
AB067488.1 |
- |
22, 23 |
2555 |
|
AF155113.1 |
- |
6, 7 |
603 |
|
AL050385.1 |
- |
23, 24 |
2549 |
|
BC114491.1 |
- |
21, 22 |
2194 |
40082305 |
CR933642.1 |
- |
24, 25 |
2683 |
|
CR936672.1 |
- |
21, 22 |
2545 |
|
View Full Product Details