Gene Transcripts

Gene Symbol
LCE2B
Entrez Gene ID
26239
Gene Name
late cornified envelope 2B
Gene Aliases
LEP10, SPRL1B, XP5
Chromosome Location
Chr. 1: 152686123 - 152687401 on Build GRCh38
UniGene ID
Hs.234766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014357.4 NP_055172.1 2 488
GenBank mRNA AF005080.1 - 2 485
AF086289.1 - 2 488
BC117235.1 - 2 479
BC117237.1 - 2 479

View Full Product Details