Gene Transcripts

Gene Symbol
Lce1e
Entrez Gene ID
68694
Gene Name
late cornified envelope 1E
Gene Aliases
1110031B11Rik, AI507275
Chromosome Location
Chr. 3: 92707400 - 92709074 on Build GRCm38
UniGene ID
Mm.290624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026811.2 NP_081087.1 2 567
GenBank mRNA AK003998.1 - 2 568
BC059085.1 - 1 526
BC099588.1 - 1 525

View Full Product Details