Gene Transcripts

Gene Symbol
Man2b2
Entrez Gene ID
360955
Gene Name
mannosidase, alpha, class 2B, member 2
Gene Aliases
Ab2-450
Chromosome Location
Chr. 14: 78939959 - 78964961 on Build Rnor_6.0
UniGene ID
Rn.139036
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134971.2 NP_001128443.1 12, 13 1990
GenBank mRNA BC168692.1 - 12, 13 1991

View Full Product Details