Gene Transcripts

Gene Symbol
PKN3
Entrez Gene ID
29941
Gene Name
protein kinase N3
Gene Aliases
UTDP4-1
Chromosome Location
Chr. 9: 128702497 - 128720920 on Build GRCh38
UniGene ID
Hs.300485
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317926.1 NP_001304855.1 20 2776
NM_013355.4 NP_037487.2 20 2776
XM_005251946.3 XP_005252003.1 20 3331
XM_006717080.2 XP_006717143.1 20 2845
XM_017014649.1 XP_016870138.1 20 2847
XM_017014650.1 XP_016870139.1 20 2985
GenBank mRNA AB019692.1 - 20 2364
BC062558.1 - 20 2379 6649806

View Full Product Details