Gene Transcripts

Gene Symbol
MRPL4
Entrez Gene ID
51073
Gene Name
mitochondrial ribosomal protein L4
Gene Aliases
CGI-28, L4mt
Chromosome Location
Chr. 19: 10251964 - 10260060 on Build GRCh38
UniGene ID
Hs.279652
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015956.2 NP_057040.2 8 999
NM_146387.1 NP_666499.1 9 863
NM_146388.1 NP_666500.1 8 999
XM_011528045.2 XP_011526347.1 8 994
GenBank mRNA AB049635.1 - 8 714
AF132962.1 - 8 960
AY007152.1 - 7 652
BC000756.2 - 8 953 2819730
BC009858.2 - 8 740 3928448
BM550378.1 - 9 861

View Full Product Details