Gene Transcripts
- Gene Symbol
- ROBO4
- Entrez Gene ID
- 54538
- Gene Name
- roundabout guidance receptor 4
- Gene Aliases
- ECSM4, MRB
- Chromosome Location
- Chr. 11: 124884218 - 124897935 on Build GRCh38
- UniGene ID
- Hs.524121
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301088.1 |
NP_001288017.1
|
9, 10 |
1441 |
|
NM_019055.5 |
NP_061928.4
|
9, 10 |
1647 |
|
XM_006718861.1 |
XP_006718924.1
|
9, 10 |
1580 |
|
XM_011542875.1 |
XP_011541177.1
|
3, 4 |
409 |
|
GenBank mRNA |
AF361473.1 |
- |
9, 10 |
1576 |
|
AK000805.1 |
- |
1, 2 |
103 |
|
AK027852.1 |
- |
9, 10 |
1546 |
|
AK074163.1 |
- |
8, 9 |
1177 |
|
AK289769.1 |
- |
9, 10 |
1546 |
|
AK297845.1 |
- |
3, 4 |
409 |
|
AK302628.1 |
- |
9, 10 |
1311 |
|
AY358083.1 |
- |
9, 10 |
1498 |
|
BC014995.1 |
- |
8, 9 |
1259 |
|
BC039602.1 |
- |
9, 10 |
1535 |
5590503 |
BC111562.1 |
- |
9, 10 |
1541 |
40068748 |
BC111748.1 |
- |
9, 10 |
1541 |
40068752 |
View Full Product Details