Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276378.1 | NP_001263307.1 | 5 | 727 | |
NM_001276379.1 | NP_001263308.1 | 4 | 602 | ||
NM_020347.3 | NP_065080.1 | 3 | 446 | ||
NR_075080.1 | 2 | 321 | |||
XM_006713207.2 | XP_006713270.2 | 3 | 431 | ||
XM_011533838.2 | XP_011532140.1 | 3 | 370 | ||
XM_017006644.1 | XP_016862133.1 | 6 | 1084 | ||
XM_017006645.1 | XP_016862134.1 | 5 | 959 | ||
GenBank mRNA | AJ297351.1 | - | 3 | 403 | |
AK024323.1 | - | 3 | 397 | ||
AK093705.1 | - | 5 | 1045 | ||
AK303416.1 | - | 6 | 498 | ||
BC025988.2 | - | 3 | 375 | 4838035 | |
BC042483.1 | - | 3 | 406 | 4831113 | |
BX640604.1 | - | 3 | 431 | ||
DA408809.1 | - | 2 | 292 | ||
HY159348.1 | - | 3 | 447 |