Gene Transcripts
- Gene Symbol
- MRPL20
- Entrez Gene ID
- 55052
- Gene Name
- mitochondrial ribosomal protein L20
- Gene Aliases
- L20mt, MRP-L20
- Chromosome Location
- Chr. 1: 1401896 - 1407313 on Build GRCh38
- UniGene ID
- Hs.182698
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318485.1 |
NP_001305414.1
|
2, 3 |
302 |
|
NM_017971.3 |
NP_060441.2
|
2, 3 |
302 |
|
GenBank mRNA |
AB049644.1 |
- |
2, 3 |
233 |
|
AK301440.1 |
- |
2, 3 |
282 |
|
AK315794.1 |
- |
2, 3 |
236 |
|
BC009515.1 |
- |
2, 3 |
270 |
3542715 |
BC014316.1 |
- |
2, 3 |
239 |
4045076 |
BC059945.1 |
- |
2, 3 |
220 |
5397911 |
BG700526.1 |
- |
2, 3 |
302 |
|
BM818788.1 |
- |
1, 2 |
99 |
|
DN990876.1 |
- |
2, 3 |
274 |
|
View Full Product Details