Gene Transcripts

Gene Symbol
LRRC36
Entrez Gene ID
55282
Gene Name
leucine rich repeat containing 36
Gene Aliases
RORBP70, XLHSRF2
Chromosome Location
Chr. 16: 67326844 - 67385206 on Build GRCh38
UniGene ID
Hs.125139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161575.1 NP_001155047.1 10, 11 1805
NM_018296.5 NP_060766.5 13, 14 2075
XM_005256025.2 XP_005256082.1 12, 13 1960
XM_005256026.2 XP_005256083.1 12, 13 1634
XM_005256027.2 XP_005256084.1 11, 12 1814
XM_005256028.1 XP_005256085.1 12, 13 1947
XM_011523201.1 XP_011521503.1 10, 11 1695
XM_011523202.2 XP_011521504.1 9, 10 1550
XM_011523203.1 XP_011521505.1 10, 11 1457
XM_011523204.1 XP_011521506.1 11, 12 1874
XM_011523205.1 XP_011521507.1 12, 13 1965
XM_017023401.1 XP_016878890.1 10, 11 1178
XM_017023402.1 XP_016878891.1 8, 9 1001
GenBank mRNA AK001866.1 - 9, 10 1708
AK292311.1 - 10, 11 1784
AK297329.1 - 7, 8 1142
AK301766.1 - 8, 9 1447
AY313780.1 - 10, 11 1768
AY963267.1 - 13, 14 2075
BC018031.1 - 12, 13 1569
BC026156.2 - 13, 14 2075 4828487

View Full Product Details