Gene Transcripts

Gene Symbol
MUC13
Entrez Gene ID
56667
Gene Name
mucin 13, cell surface associated
Gene Aliases
DRCC1, MUC-13
Chromosome Location
Chr. 3: 124905442 - 124934751 on Build GRCh38
UniGene ID
Hs.5940
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033049.3 NP_149038.3 3 590
GenBank mRNA AB035807.1 - 3 575
AF286113.1 - 3 578
AY358831.1 - 3 575

View Full Product Details