Gene Transcripts
- Gene Symbol
- MRPS15
- Entrez Gene ID
- 64960
- Gene Name
- mitochondrial ribosomal protein S15
- Gene Aliases
- DC37, MPR-S15, RPMS15, S15mt
- Chromosome Location
- Chr. 1: 36455761 - 36464439 on Build GRCh38
- UniGene ID
- Hs.352839
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031280.3 |
NP_112570.2
|
3, 4 |
424 |
|
GenBank mRNA |
AB049946.1 |
- |
3, 4 |
299 |
|
AB209905.1 |
- |
3, 4 |
323 |
|
AF265439.1 |
- |
3, 4 |
343 |
|
AK302633.1 |
- |
3, 4 |
368 |
|
AK315441.1 |
- |
3, 4 |
363 |
|
BC031336.1 |
- |
3, 4 |
339 |
3543896 |
BF310592.1 |
- |
3, 4 |
357 |
|
BM048518.1 |
- |
3, 4 |
423 |
|
BU927793.1 |
- |
3, 4 |
274 |
|
View Full Product Details