Gene Transcripts

Gene Symbol
OXER1
Entrez Gene ID
165140
Gene Name
oxoeicosanoid receptor 1
Gene Aliases
GPCR, GPR170, TG1019
Chromosome Location
Chr. 2: 42762499 - 42764261 on Build GRCh38
UniGene ID
Hs.168439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148962.4 NP_683765.1 1 128
GenBank mRNA AB083055.2 - 1 84
BC063549.1 - 1 128 4777971

View Full Product Details