Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017924.3 | NP_001017924.1 | 3 | 1157 | |
NM_001284223.1 | NP_001271152.1 | 4 | 1187 | ||
GenBank mRNA | BC140765.1 | - | 4 | 1187 | |
BC144475.1 | - | 4 | 1187 | 9053002 | |
BX648524.1 | - | 3 | 1097 |