Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr. 14: 88824153 - 88877996 on Build GRCh38
UniGene ID
Hs.303055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288781.1 NP_001275710.1 14, 15 1600
NM_001288782.1 NP_001275711.1 12, 13 1520
NM_001288783.1 NP_001275712.1 13, 14 1492
NM_144596.3 NP_653197.2 14, 15 1552
NM_198309.3 NP_938051.1 13, 14 1522
NM_198310.3 NP_938052.1 12, 13 1432
XM_006720035.1 XP_006720098.1 12, 13 1804
XM_006720037.2 XP_006720100.1 11, 12 1714
XM_011536432.1 XP_011534734.1 13, 14 1882
XM_011536434.1 XP_011534736.1 12, 13 1792
XM_011536435.2 XP_011534737.1 12, 13 1268
GenBank mRNA AK093891.1 - 12, 13 1370
AK124675.1 - 13, 14 1402
AK315772.1 - 12, 13 1407
AL833901.1 - 11, 12 1256
AY366523.1 - 12, 13 1378
AY366524.1 - 11, 12 1288
AY373972.1 - 12, 13 1378
BC001563.1 - 8, 9 870
BC026351.1 - 13, 14 1456 4799330
BC095433.1 - 12, 13 1360 30368216
BX161472.1 - 11, 12 1360
BX247959.1 - 12, 13 1469
BX248071.1 - 4, 5 609
BX248248.1 - 9, 10 964
BX537546.1 - 1 1096
KU178849.1 - 13, 14 1404
KU178850.1 - 12, 13 1326
KU178851.1 - 11, 12 1236
KU178853.1 - 6, 7 671

View Full Product Details