Gene Transcripts

Gene Symbol
Nos3
Entrez Gene ID
18127
Gene Name
nitric oxide synthase 3, endothelial cell
Gene Aliases
2310065A03Rik, Nos-3, eNOS, ecNOS
Chromosome Location
Chr. 5: 24364819 - 24384474 on Build GRCm38
UniGene ID
Mm.258415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008713.4 NP_032739.3 21 2838
GenBank mRNA AK077896.1 - 21 2840
AK143221.1 - 20 2610
AK158106.1 - 21 2860
BC052636.1 - 21 2825 6530765
U53142.1 - 21 2826

View Full Product Details