Gene Transcripts

Gene Symbol
Olfr65
Entrez Gene ID
18365
Gene Name
olfactory receptor 65
Gene Aliases
5'[b]3, 5'beta3, MOR1-1, MOR1-2, ORL670, Olfr64
Chromosome Location
Chr. 7: 103906342 - 103907447 on Build GRCm38
UniGene ID
Mm.377106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013616.4 NP_038644.2 1 723
GenBank mRNA BC137766.1 - 1 723

View Full Product Details