Gene Transcripts

Gene Symbol
Rad21
Entrez Gene ID
19357
Gene Name
RAD21 homolog (S. pombe)
Gene Aliases
SCC1, mKIAA0078
Chromosome Location
Chr. 15: 51962604 - 51993538 on Build GRCm38
UniGene ID
Mm.182628
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009009.4 NP_033035.3 9 1266
GenBank mRNA AF332085.1 - 8 1021
AF332086.1 - 8 1021
AK004746.1 - 9 1069
AK128984.1 - 9 1155
AK129050.1 - 4 394
AK139492.1 - 8 919
AK145591.1 - 9 1233
AK163992.1 - 1127
AK168718.1 - 9 1254
AK168896.1 - 9 1215
BC043032.1 - 8 1035 5696540
BC129916.1 - 8 1065 40111820
BC129917.1 - 8 1066 40111821
D37790.1 - 8 953
X98293.1 - 8 1021

View Full Product Details