Gene Transcripts

Gene Symbol
Mst1r
Entrez Gene ID
19882
Gene Name
macrophage stimulating 1 receptor (c-met-related tyrosine kinase)
Gene Aliases
CD136, CDw136, Fv-2, Fv2, PTK8, Ron, STK
Chromosome Location
Chr. 9: 107906860 - 107920383 on Build GRCm38
UniGene ID
Mm.3901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009074.2 NP_033100.2 1 922
GenBank mRNA AK053655.1 - 1 905
BC141379.1 - 1 733 9055992
X74736.1 - 1 922

View Full Product Details