Gene Transcripts

Gene Symbol
Morf4l1
Entrez Gene ID
21761
Gene Name
mortality factor 4 like 1
Gene Aliases
MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
Chromosome Location
Chr. 9: 90091669 - 90114820 on Build GRCm38
UniGene ID
Mm.426209
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039147.2 NP_001034236.1 11 1029
NM_024431.3 NP_077751.1 10 912
GenBank mRNA AB042241.1 - 10 761
AF319620.1 - 10 760
AF319621.1 - 11 877
AK010528.1 - 10 680
AK011249.1 - 10 789
AK011441.1 - 10 684
AK016414.1 - 10 879
AK041833.1 - 2 1223
AK079115.1 - 6 362
AK146210.1 - 10 747
AK146785.1 - 9 642
AK150925.1 - 10 713
AK153545.1 - 5 345
AK167079.1 - 10 671
AK168327.1 - 10 747
AK168585.1 - 10 692
AK168608.1 - 10 694
AK168674.1 - 10 743
AK169065.1 - 10 744
AK184930.1 - 4 253
AK205535.1 - 4 253
AK213016.1 - 4 253
AK214961.1 - 4 254
AK216220.1 - 4 253
AK216852.1 - 3 254
AK220441.1 - 8 3688
BC003894.1 - 10 764 3593982
BC017619.1 - 10 764 4923923
BC083094.1 - 10 698
BC083118.1 - 10 773 5371036
BC085103.1 - 11 879 6490291
BC092293.1 - 10 776 5068516
BC103783.1 - 10 776 6529024

View Full Product Details