Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024193.2 | NP_077155.2 | 9 | 1200 | |
GenBank mRNA | AK009799.1 | - | 9 | 1186 | |
AK011744.1 | - | 9 | 1164 | ||
AK013528.1 | - | 9 | 1161 | ||
AK030730.1 | - | 5 | 1919 | ||
AK037405.1 | - | 1 | 186 | ||
AK077795.1 | - | 9 | 1167 | ||
AK139924.1 | - | 9 | 1167 | ||
AK150258.1 | - | 9 | 1167 | ||
AK154024.1 | - | 6 | 1767 | ||
BC002231.1 | - | 4 | 552 | ||
BC021355.1 | - | 9 | 1162 | 5006404 |