Gene Transcripts

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
Chromosome Location
Chr. 14: 23412738 - 23435686 on Build GRCh38
UniGene ID
Hs.719946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000257.3 NP_000248.2 16 1947
XM_017021340.1 XP_016876829.1 15 1882
GenBank mRNA AB209708.1 - 5 734
AK307590.1 - 16 1921
BC112171.1 - 15 1871 8327639
BC112173.1 - 15 1871 8327641
EF560725.1 - 14 1814
EU747717.1 - 16 1922
M58018.1 - 16 1902

View Full Product Details