Gene Transcripts

Gene Symbol
Lztfl1
Entrez Gene ID
93730
Gene Name
leucine zipper transcription factor-like 1
Gene Aliases
5530402H04Rik, 6130400H19Rik, AI414725, AW048545
Chromosome Location
Chr. 9: 123694413 - 123717668 on Build GRCm38
UniGene ID
Mm.378263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033322.2 NP_201579.1 2 91
GenBank mRNA AJ297352.1 - 2 87
AK029671.1 - 2 123
AK030068.1 - 2 103
AK041091.1 - 2 124
AK046515.1 - 2 80
AK054003.1 - 2 97
AK145263.1 - 2 100
AK157770.1 - 2 98
BC026840.1 - 2 91 4165152
BC037045.1 - 2 91
BC098224.1 - 2 96 1263280

View Full Product Details