Gene Transcripts

Gene Symbol
Adgrv1
Entrez Gene ID
110789
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
Frings, Gpr98, Mass1, Mgr1, VLGR1
Chromosome Location
Chr. 13: 81095068 - 81633197 on Build GRCm38
UniGene ID
Mm.288694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054053.4 NP_473394.3 9 1659
GenBank mRNA AB086166.2 - 9 1654
AB086167.1 - 9 1654
AF405692.1 - 5 1804
AF435926.1 - 9 1660

View Full Product Details