Gene Transcripts

Gene Symbol
Olfr53
Entrez Gene ID
258962
Gene Name
olfactory receptor 53
Gene Aliases
IE12, MOR253-5
Chromosome Location
Chr. 7: 140646452 - 140652919 on Build GRCm38
UniGene ID
Mm.246542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146960.2 NP_667171.1 2 1063
GenBank mRNA BC119325.1 - 1 863
BC120694.1 - 1 865 40129617

View Full Product Details