Gene Transcripts

Gene Symbol
STK16
Entrez Gene ID
8576
Gene Name
serine/threonine kinase 16
Gene Aliases
KRCT, MPSK, PKL12, TSF1
Chromosome Location
Chr. 2: 219245469 - 219250337 on Build GRCh38
UniGene ID
Hs.153003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008910.2 NP_001008910.1 4 507
NR_026909.1 4 772
XM_011512067.2 XP_011510369.1 4 508
GenBank mRNA AB020739.1 - 3 401
AF060798.1 - 4 478
AF203910.1 - 4 474
AJ005791.1 - 3 468
AJ010872.1 - 3 378
AK225392.1 - 4 472
AK225526.1 - 479
AK292694.1 - 4 507
AK302182.1 - 4 741
AK310793.1 - 4 772
AL552406.3 - 4 749
BC002618.2 - 4 476 3143105
BC053998.1 - 4 472 5760866
BT019585.1 - 3 350
CR407675.1 - 3 350

View Full Product Details